Learn More
Selleck Chemical LLCSupplier Diversity Partner Antazoline HCl DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S419925MG
Antazoline HCl is a first generation antihistamine, binding to the histamine H1 receptor and blocking the action of endogenous histamine.
Catalog No. 50-194-8422