Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene, 97%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
120-12-7
120-12-7
Packaging Poly Bottle
Quantity 25 g
CAS Max % 1
Percent Purity 0.97
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.