Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anthracene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Anthracene is used in the production of the red dyealizarin and other dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Anthracene is used in the production of the red dyealizarin and other dyes.
Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Anthracene is used in the production of the red dyealizarin and other dyes.
Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
Specifications
Melting Point | 214°C to 218°C |
Density | 1.25 |
Boiling Point | 340°C to 342°C |
Flash Point | 121°C (249°F) |
Quantity | 500 g |
UN Number | UN3077 |
Beilstein | 1905429 |
Merck Index | 14,682 |
Solubility Information | Soluble in alcohol,methanol (1.8g/100 g),hexane (0.37g/100 g). Insoluble in water. |
Formula Weight | 178.23 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H500 |
Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 204-371-1 |
RTECSNumber | CA9350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction