Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene, Practical, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
120-12-7
120-12-7
Packaging Poly Pail
Melting Point 215° to 219°C
Quantity 10 kg
CAS Max % 1
Grade Practical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.