Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthracene 96.0+%, TCI America™

Click to view available options
Quantity:
25g
120-12-7
C14H10
178.234
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene,anthracin,green oil,anthracen,tetra olive n2g,anthracen german,anthrazen,anthracene, pure,bis-alkylamino,ccris 767
8418
CHEBI:35298
anthracene
C1=CC=C2C=C3C=CC=CC3=CC2=C1
120-12-7
C14H10
178.234
MFCD00001240
MWPLVEDNUUSJAV-UHFFFAOYSA-N
paranaphthalene,anthracin,green oil,anthracen,tetra olive n2g,anthracen german,anthrazen,anthracene, pure,bis-alkylamino,ccris 767
8418
CHEBI:35298
anthracene
C1=CC=C2C=C3C=CC=CC3=CC2=C1
Color White-Yellow
Boiling Point 342°C
Melting Point 217°C
Quantity 25g
UN Number 3077
Formula Weight 178.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material Anthracene

EINECSNumber : (4)-0683

RTECSNumber : CA9350000

TSCA : Yes