Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthraquinone-1,5-disulfonic Acid Disodium Salt 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
853-35-0
C14H6Na2O8S2
412.294
MFCD00044659
OFASSSMJNCWWTP-UHFFFAOYSA-L
anthraquinone-1,5-disulfonic acid disodium salt,1,5-anthraquinonedisulfonic acid, disodium salt,sodium anthraquinone-1,5-disulfonate,disodium anthraquinone-1,5-disulfonate,1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, disodium salt,anthrachinon-1,5-disulfonan sodny czech,anthrachinon-1,5-disulfonan disodny czech,disodium 9,10-dihydro-9,10-dioxoanthracene-1,5-disulphonate,1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt 1:2,1,5-bis sodiooxysulfonyl-9,10-anthraquinone
70067
disodium;9,10-dioxoanthracene-1,5-disulfonate
C1=CC2=C(C(=C1)S(=O)(=O)[O-])C(=O)C3=C(C2=O)C(=CC=C3)S(=O)(=O)[O-].[Na+].[Na+]
853-35-0
C14H6Na2O8S2
412.294
MFCD00044659
OFASSSMJNCWWTP-UHFFFAOYSA-L
anthraquinone-1,5-disulfonic acid disodium salt,1,5-anthraquinonedisulfonic acid, disodium salt,sodium anthraquinone-1,5-disulfonate,disodium anthraquinone-1,5-disulfonate,1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, disodium salt,anthrachinon-1,5-disulfonan sodny czech,anthrachinon-1,5-disulfonan disodny czech,disodium 9,10-dihydro-9,10-dioxoanthracene-1,5-disulphonate,1,5-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt 1:2,1,5-bis sodiooxysulfonyl-9,10-anthraquinone
70067
disodium;9,10-dioxoanthracene-1,5-disulfonate
C1=CC2=C(C(=C1)S(=O)(=O)[O-])C(=O)C3=C(C2=O)C(=CC=C3)S(=O)(=O)[O-].[Na+].[Na+]
Color Undesignated
Quantity 500g
Formula Weight 412.29
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Anthraquinone-1,5-disulfonic Acid Disodium Salt

EINECSNumber : (4)-0725

RTECSNumber : CB0555500

TSCA : Yes