Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthraquinone-1-diazonium Chloride, MP Biomedicals

Click to view available options
Quantity:
1g
16048-40-1
C14H7N2O2+
235.222
AYWYBCRYWKVVBR-UHFFFAOYSA-N
1-anthracenediazonium, 9,10-dihydro-9,10-dioxo, 9,10-dihydro-9,10-dioxoanthracenediazonium, 1-aminoanthraquinone diazonium salt
27658
9,10-dioxoanthracene-1-diazonium
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+]#N
16048-40-1
C14H7N2O2+
235.222
AYWYBCRYWKVVBR-UHFFFAOYSA-N
1-anthracenediazonium, 9,10-dihydro-9,10-dioxo, 9,10-dihydro-9,10-dioxoanthracenediazonium, 1-aminoanthraquinone diazonium salt
27658
9,10-dioxoanthracene-1-diazonium
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C(=CC=C3)[N+]#N
Quantity 1g
Formula Weight 16048-40-1
Chemical Name or Material Anthraquinone-1-diazonium Chloride