Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anthrax PA Mouse anti-Bacteria, Clone: PA6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP1975350.025MG
Description
Anthrax PA Monoclonal specifically detects Anthrax PA in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Anthrax PA | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
anthrax PA, Anthrax toxins translocating protein, Bacillus anthracis protective antigen, PA, PA 83, PA83, PAG, Paga, Protective antigen | |
Anthrax protective antigen. | |
0.025 mg | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunoprecipitation, Western Blot | |
PA6 | |
Unconjugated | |
PBS with 0.01% Sodium Azide | |
Mouse | |
Protein G purified | |
RUO | |
Recognizes Bacteria Anthrax Protective Antigen (PA). | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only