Learn More
Anthrone, 95%, Thermo Scientific Chemicals
Description
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 153°C to 160°C |
Color | Yellow |
Quantity | 25 g |
Beilstein | 1910173 |
Sensitivity | Light sensitive |
Merck Index | 14,691 |
Solubility Information | Soluble in alcohol, benzene, and hot NaOH solutions; Insoluble in water |
IUPAC Name | 10H-anthracen-9-one |
Formula Weight | 194.23 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 201-994-0 |
RTECSNumber | CB8925500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.