Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 eta Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528113

Catalog No. PIPA528113


Add to cart

Description

Description

PA5-28113 targets 14-3-3 eta in WB applications and shows reactivity with Human samples. The PA5-28113 immunogen is synthetic peptide corresponding to a region within amino acids 26 and 120 of Human 14-3-3 eta. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and bovine orthologs. This gene contains a 7 bp repeat sequence in its 5 ft. UTR, and changes in the number of this repeat has been associated with early-onset schizophrenia.
Specifications

Specifications

14-3-3 eta
Polyclonal
Unconjugated
YWHAH
Liquid
YWHAH
Synthetic peptide corresponding to a region within amino acids 26 and 120 of Human 14-3-3 eta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q04917
YWHA1, 14-3-3 eta, 14-3-3 protein eta
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7533
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit