Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 beta Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137002

Catalog No. 01-673-011


Add to cart

Description

Description

This antibody is predicted to react with rat and mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain tissue.

At least 7 mammalian isoforms comprise the 14-3-3 family of signaling intermediates. 14-3-3 molecules are highly conserved, expressed in a broad range of tissues and cell types and were first discovered as a series of highly abundant proteins in brain tissue. There is increasing evidence that 14-3-3 proteins are critical to cell transformation and mitotic signaling.
Specifications

Specifications

14-3-3 beta
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P31946, Q9CQV8
GW128, HS1, KCIP1
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
54401, 7529
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
YWHAB
Liquid
YWHAB
Synthetic peptide from N-terminus of human 14-3-3 beta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit