Learn More
14-3-3 ζ/γ/η Rabbit anti-Vertebrates, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06408
Catalog No. 06-408-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect 14-3-3 ζ/γ/η using this Anti-14-3-3 ζ/γ/η Antibody validated for use in western blotting.
Specifications
Specifications
14-3-3 ζ/γ/η
Unconjugated
Serum
Rabbit
IgG
150μL
Primary
NM_003406
Western Blot
rabbit antiserum
YWHAZ; KCIP-1; MGC126532; MGC111427; MGC138156
HPLC-purified synthetic peptide (KMKGDYYRYLAE) corresponding to the zeta, gamma and eta isoforms of 14-3-3
Antiserum
−20°C for 24 months
Polyclonal
Vertebrate