Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 zeta Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515471

Catalog No. PIPA515471

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken and rat based on sequence homology.

YWHAZ belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Two transcript variants differing in the 5 ft. UTR, but encoding the same protein, have been identified for the gene. Both variants encode the same protein, however, they are differentially expressed in hematopoietic cells.
Specifications

Specifications

14-3-3 zeta
Polyclonal
PBS with 0.09% sodium azide
P63104, P63101
14-3-3 protein zeta/delta; Protein kinase C inhibitor protein-1; KCIP-1; Factor activating exoenzyme S; FAS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22631, 7534
Immunohistochemistry, Western Blot
Unconjugated
YWHAZ
Liquid
YWHAZ
KLH conjugated synthetic peptide between 216-245 amino acids from human YWHAZ
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit