Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 sigma Rabbit anti-Amphibian, Bovine, Canine, Equine, Human, Marsupial, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523507

Catalog No. PIPA523507


Add to cart

Description

Description

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigma is the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to g-irradiation and other DNA-damaging agents. 14-3-3 sigma is a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5 ft. CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
Specifications

Specifications

14-3-3 sigma
Polyclonal
Unconjugated
SFN
Liquid
SFN
Synthetic peptide (133 EVATGDDKKRIIDSARSA 150) of 14-3-3 sigma protein.
Protein A
RUO
Antibody
Polyclonal
Amphibian, Bovine, Canine, Equine, Human, Marsupial, Non-human Primate
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide
Q0VC36, P31947
YWHAS, 14-3-3 protein sigma, 14-3-3 sigma, epithelial cell marker protein 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100057082, 2810, 487351, 528453
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit