Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 zeta Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137006

Catalog No. 01-673-012

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken, mouse and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil or breast carcinoma.

14-3-3 proteins are present as multigene families in most organisms. Signal-induced phosphorylation has the ability to change protein function. Sometimes, however, phosphorylation is not enough to change a protein’s function. 14-3-3 proteins play an important role of bringing signal transduction to completion. They regulate many cellular processes that are important in cancer biology, such as apoptosis and cell-cycle. checkpoints.
Specifications

Specifications

14-3-3 zeta
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P63104
Protein kinase C inhibitor protein 1, KCIP-1
Rabbit
IgG
1 mL
4° C, do not freeze
Primary
7534
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
YWHAZ
Liquid
YWHAZ
Synthetic peptide derived from internal region of human 14-3-3 zeta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit