Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-PGDH Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522889

Catalog No. PIPA522889


Add to cart

Description

Description

Suggested positive control: antigen standard for HPGD (transient overexpression lysate), Treated A549 lung carcinoma cell lysates and LoVo colon carcinoma cell lysates.

15-Hydroxyprostaglandin dehydrogenase (15-PGDH) is a novel tumor suppressor in the COX-2 pathway. The protein is found in many normal tissues and appears to be down-regulated in colorectal and lung carcinoma tissue.
Specifications

Specifications

15-PGDH
Polyclonal
Biotin
HPGD
Liquid
HPGD
Purified type I human placental 15-PGDH protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1.9 mg/mL
PBS with 0.05% sodium azide
P15428, Q8VCC1, O08699
15-PGDH, PGDH, PGDH1, PHOAR1, SDR36C1, 15-hydroxyprostaglandin dehydrogenase [NAD+], NAD+-dependent 15-hydroxyprostaglandin dehydrogenase, prostaglandin dehydrogenase 1, short chain dehydrogenase/reductase family 36C, member 1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
15446, 3248, 79242
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit