Learn More
MilliporeSigma™ anti-20S Proteasome alpha1, 2, 3, 5, 6, 7-Subunits Clone: MCP231, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1049100UL
Catalog No. ST1049100UL
Add to cart
Description
Description
Specifically detects 20S Proteasome α1, 2, 3, 5, 6, & 7-Subunits Clone: MCP231 in Human, Mouse, Rabbit, Rat, Yeast samples, and it is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immuno Blotting
Proteasomes are large multi-subunit complexes, that are localized in the nucleus and cytosol, and selectively degrade intracellular proteins. They play a major role in the degradation of many proteins involved in cellular processes such as cell cycling, apoptosis, and proliferation. The human proteasome contains two copies of each of seven different α-type and β-type subunits assembled to form a stack of four seven-membered rings. The α-subunits comprise the outer rings while the β-subunits comprise the inner rings. The α-subunits contain information for the binding of regulatory complexes such as PA700 and PA28.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
20S Proteasome α1, 2, 3, 5, 6, & 7-Subunits
MCP231
In PBS.
Murine
IgG1
RUO
Primary
Recognizes α1, 2, 3, 5, 6 and 7 subunits of 20S proteasome in human placental proteasome preparations.
ELISA, Immuno Blotting, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Unconjugated
Purified
dinitrophenylated proteasomes
100μL
−20°C
Monoclonal
Human, Murine, Rabbit, Rat, Yeast