Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

septin 4, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human SEPT4 protein.

Manufacturer:  Abnova Corporation H00005414B01

Catalog No. 89-005-981


Add to cart

Description

Description

This gene is a member of the septin gene family of nucleotide binding proteins, originally described in yeast as cell division cycle regulatory proteins. Septins are highly conserved in yeast, Drosophila, and mouse and appear to regulate cytoskeletal organization. The protein encoded by this gene is thought to be part of a complex involved in cytokinesis. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq

Sequence: MDRSLGWQGNSVPEDRTEAGIKRFLEDTTDDGELSKFVKDFSGNASCHPPEAKTWASRPQVPEPRPQAPDLYDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKSVKKGFDFTLMVAGESGLGKSTLVNSLFLTDLYRDRKLLGAEERIMQTVEITKHAVDIEEKGVRLRLTIVDTPGFGDAVNNTECWKPVAEYIDQQFEQYFRDESGLNRKNIQDNRVHCCLYFISPFGHGLRPLDVEFMKALHQRVNIVPILAKADTLTPPEVDHKKRKIREEIEHFGIKIYQFPDCDSDEDEDFKLQDQALKESIPFAVIGSNTVVEARGRRVRGRLYPWGIVEVENPGHCDFVKLRTMLVRTHMQDLKDVTRETHYENYRAQCIQSMTRLVVKERNRNKLTRESGTDFPIPAVPPGTDPETEKLIREKDEELRRMQEMLHKIQKQMKENY
Specifications

Specifications

Septin 4
Unconjugated
No additive
NM_004574
ARTS|BRADEION|CE5B3|H5|MART|PNUTL2|SEP4|hCDCREL-2|hucep-7
Mouse
50 uL
Apoptosis
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human SEPT4 protein.
SEPT4
Serum
SEPT4
SEPT4 (NP_004565, 1 a.a. ~ 478 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5414
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit