Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR3A Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130207

Catalog No. PIPA130207


Add to cart

Description

Description

PA1-30207 detects 5-HT3 receptor from rat, human samples. PA1-30207 has been successfully used in Western blot applications. A suggested positive control for this product is human brain tissue. The PA1-30207 immunogen is synthetic peptides, corresponding to amino acids 21-36 and 421-436 of Rat 5HT3 Receptor. Store at 4°C short term. For extended storage, store at -20°C, avoiding freeze/thaw cycles.

receptor for 5-hydroxytryptamine (serotonin); functions as a ligand-gated ion channel.
Specifications

Specifications

HTR3A
Polyclonal
Unconjugated
HTR3A
Liquid
HTR3A
Synthetic peptides, corresponding to amino acids 21-36 and 421-436 of Rat 5HT3 Receptor.
Protein G
RUO
Antibody
Polyclonal
Human, Rat
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.05% sodium azide
P46098, P35563
5-HT3, 5-HT3A, 5HT3, Htr3
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3359, 79246
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit