Learn More
5-Hydroxymethyluridine Goat anti-Human, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen PA186851
Catalog No. PIPA186851
Add to cart
Description
Description
5-hydroxymethyluridine is created by the oxidation of thymine by ionising radiation or reactive oxygen species. This oxidation causes changes in gene expression, mutations and chromosomal instability all of which play a role in cancer, autoimmune disease and aging. 5-hydroxymethyluridine is a potential biomarker for female breast, colon and rectal cancers, being present at elevated levels, possibly due to impaired DNA repair mechanisms.
Specifications
Specifications
5-Hydroxymethyluridine
Polyclonal
whole serum with 0.1% sodium azide
5-(Hydroxymethyl)uridine; C10H14N2O7
Synthetic Peptide corresponding to 5-hydroxymethyluridine
100 μL
4°C or -20°C if preferred
Primary
Human
ELISA
Unconjugated
Liquid
Goat
IgG
RUO
Antibody
Polyclonal