Learn More
53BP1 Rabbit anti-Human, Mouse, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522790
Catalog No. PIPA522790
Add to cart
Description
Description
This antibody reacts with Human samples in Western blot and ICC applications, and with Mouse in ICC applications.
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Specifications
Specifications
53BP1
Polyclonal
Biotin
TP53BP1
Liquid
TP53BP1, Trp53bp1
A synthetic peptide, which represented a portion of human tumor protein p53-binding protein 1 encoded in exon 11, 12 and 19 (LocusLink ID 7158). This antibody is a cocktail hybrid of NB 100-304 and NB 100-305.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with no preservative
Q12888, P70399
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
27223, 7158