Learn More
MilliporeSigma™ anti-53BP1, Clone: BP13, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB3802/DEL
Encompass
Catalog No. 50-175-402
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
P53-binding protein 1 (53BP1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, it is phosphorylated and relocalizes to the presumptive sites of damage, specifically, double-strand breaks. This also suggests a role in DNA repair, and maintaining genomic stability. It is sequence specific transcription factor that plays a central role in the the response of mammalian cells to genotoxic stress. Also, 53BP1 localizes to kinetochores in mitotic cells, suggesting a potential function in mitotic checkpoint signaling.
Immunofluorescence, Western Blotting
Specifications
Specifications
53BP1
Unconjugated
Recombinant human 53BP1 proteins
RUO
Primary
Semi-purified
Monoclonal
Mouse
100 μg
Epigenetics
Human
IgG1
Product Suggestions
Product Suggestions