Learn More
53BP1 Rabbit anti-Human, Mouse, Rat, DyLight® 550, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522761
Catalog No. PIPA522761
Add to cart
Description
Description
Human samples have been tested in Western Blot and ICC/IF, and mouse samples have been tested in ICC/IF only. Suggested positive control: U205, 293T, and MO59K cell lysates and MEF lysates .
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Specifications
Specifications
53BP1
Polyclonal
DyLight™ 550
TP53BP1
Liquid
TP53BP1, Trp53bp1
The epitope recognized by this antibody maps to a region between residues 1925 and the C-terminus (residue 1972) of human tumor protein p53 binding protein 1 using the numbering given in entry NP_005648.1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with no preservative
Q12888, P70399
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
27223, 296099, 7158