Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
53BP1 Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522758
Description
Human samples have been tested in Western Blot and ICC/IF, and mouse samples have been tested in ICC/IF only. Suggested positive control: U205, 293T, and MO59K cell lysates and MEF lysates .
53BP1 Polyclonal antibody specifically detects 53BP1 in Bat, Bovine, Canine, Human, Mouse, Rat, Rodent samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
53BP1 | |
Polyclonal | |
DyLight 650 | |
TP53BP1 | |
53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27223, 296099, 478274, 509111, 7158 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P70399, Q12888 | |
TP53BP1, Trp53bp1 | |
The epitope recognized by this antibody maps to a region between residues 1925 and the C-terminus (residue 1972) of human tumor protein p53 binding protein 1 using the numbering given in entry NP_005648.1. | |
100 μL | |
Primary | |
Bat, Bovine, Canine, Human, Mouse, Rat, Rodent | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction