Learn More
53BP1 Rabbit anti-Human, Mouse, Rat, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522760
Catalog No. PIPA522760
Add to cart
Description
Description
Human samples have been tested in Western Blot and ICC, and mouse samples have been tested in ICC only.
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
Specifications
Specifications
53BP1
Polyclonal
HRP
TP53BP1
Liquid
TP53BP1, Trp53bp1
The epitope recognized by this antibody maps to a region between residues 1925 and the C-terminus (residue 1972) of human tumor protein p53 binding protein 1 using the numbering given in entry NP_005648.1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with no preservative
Q12888, P70399
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Rabbit
IgG
100 μL
4° C, store in dark
Primary
27223, 296099, 7158