Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP2 Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611354

Catalog No. BDB611354


Add to cart

Description

Description

The p53 protein is critical to regulation of normal cell growth and is a suppressor of tumor cell proliferation. Inactivation of p53 by a number of mechanisms, such as missense mutations or interaction with oncogenic viral or cellular proteins, can result in tumor progression. In addition, Bcl2 and p53 are involved in apoptosis in an antagonistic fashion such that overexpressed Bcl2 inhibits p53-induced apoptosis. 53BP1 and 53BP2 were identified in a yeast two-hybrid screen of proteins that bind p53. Both 53BP1 and 53BP2 bind wild type p53, but not mutant p53 found in tumor cells. p53BP1 is localized to the cytoplasm and nucleus, while p53BP2 is found only in the cytoplasm. 53BP1 has BRCT motifs found in proteins involved in cell cycle control and DNA repair. DNA damage leads to 53BP1 hyperphosphorylation, which may be mediated by ATM. 53BP2 has four ankyrin repeats and a SH3 domain that are required for interactions with Bcl2 and p53. Overexpression of 53BP2 in 293 cells inhibits progression of the cell cycle in G2/M phase, while co-transfection of 53BP2 with p53 in H358 cells enhances p53-mediated transcriptional activation. The interaction between 53BP2 and p53 may be regulated by Bcl2, since competition experiments demonstrate that Bcl2 prevents p53 binding to 53BP2. In addition, 53BP2 can also bind the apoptotic-related p65 subunit of NFκB and this subunit can inhibit 53BP2-induced cell death.

Specifications

Specifications

53BP2
19
Unconjugated
Affinity Purified
Human 53BP2 aa. 674-793
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit