Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HTR2C Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527164

Catalog No. PIPA527164


Add to cart

Description

Description

PA5-27164 targets 5HT2C Receptor in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27164 immunogen is a synthetic peptide corresponding to a region within amino acids 387 and 458 of 5HT2C Receptor. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Serotonin (5-hydroxytryptamine, 5-HT), a neurotransmitter, elicits a wide array of physiological effects by binding to several receptor subtypes, including the 5-HT2 family of seven-transmembrane-spanning, G-protein-coupled receptors, which activate phospholipase C and D signaling pathways. This gene encodes the 2C subtype of serotonin receptor and its mRNA is subject to multiple RNA editing events, where genomically encoded adenosine residues are converted to inosines. RNA editing is predicted to alter amino acids within the second intracellular loop of the 5-HT2C receptor and generate receptor isoforms that differ in their ability to interact with G proteins and the activation of phospholipase C and D signaling cascades, thus modulating serotonergic neurotransmission in the central nervous system. Studies in humans have reported abnormalities in patterns of 5-HT2C editing in depressed suicide victims.
Specifications

Specifications

HTR2C
Polyclonal
Unconjugated
HTR2C
Liquid
HTR2C
Synthetic peptide corresponding to a region within amino acids 387 and 458 of 5HT2C Receptor
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.68 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P28335
5-HT2C, 5-HTR2C, HTR1C, 5-HT-1C, 5-HT-2C, 5-HT1C, 5-hydroxytryptamine receptor 1C, 5-hydroxytryptamine receptor 2C, serotonin 5-HT-2C receptor, serotonin receptor 2C
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3358
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit