Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NT5C1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526912

Catalog No. PIPA526912

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Cytosolic nucleotidases, such as NT5C1A, dephosphorylate nucleoside monophosphates.
Specifications

Specifications

NT5C1A
Polyclonal
PBS with 0.09% sodium azide
Q9BXI3, A3KFX0
CN-I, CN-IA, CN1, CN1A, AMP-specific 5′-NT, cytosolic 5′ nucleotidase, type 1A, cytosolic 5′-nucleotidase 1A, cytosolic 5′-nucleotidase IA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
230718, 84618
Immunohistochemistry, Western Blot
Unconjugated
NT5C1A
Liquid
NT5C1A
KLH conjugated synthetic peptide between 91-119 amino acids from the N-terminal region of human 5NT1A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit