Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-6-Histidine Epitope Tag, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP125939

Catalog No. NBP125939


Add to cart

Description

Description

His Tag Polyclonal antibody specifically detects His Tag in Non-species specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

His Tag
1.0 mg/mL
Western Blot 1:1000-1:10000, ELISA 1:1000 - 1:30000, Immunocytochemistry/Immunofluorescence 1:100-1:400, Immunoprecipitation 1 - 4 ug/mg lysate
Affinity Purified
Goat
IgG
0.1 ml
Epitope Tags, His Epitope Tags
Primary
This antibody recognizes the motif H-H-H-H-H-H and is therefore of value in detecting proteins containing histidine tags. This antibody may not detect all N-terminal (NT) histidine tagged proteins.
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine
Keyhole limpet haemocyanin (KLH) conjugated H-H-H-H-H-H
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Non-species specific
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only