Learn More
6x-His Tag Rabbit anti-Tag, HRP, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA1983BHRP
Catalog No. PA1983BHRP
Add to cart
Description
Description
PA1-983B-HRP detects His-tagged proteins in Western Blot applications.
The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombit proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombit protein using genetic engineering. By choosing a particular epitope and recombit protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombit proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
Specifications
Specifications
6x-His Tag
Polyclonal
HRP
Liquid
Rabbit
IgG
50 μL
4° C, do not freeze
Primary
Tag
Western Blot
1 mg/mL
PBS with proprietary stabilizer and 0.07% Kathon
His Tag; Histidine Tag; Histidine Epitope Tag; 6X His eptope tag; 6His tag; 6X His
Synthetic peptide corresponding to residues H H H H H H
Affinity chromatography
RUO
Antibody
Polyclonal