Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-6x-His Epitope Tag Oligoclonal, Invitrogen™

6x-His Epitope TagOligoclonal antibody 710286 from Thermo Scientific™ Pierce™ specifically detects 6x-His Epitope Tag in Tag samples, and it is validated for WB

Manufacturer:  Invitrogen 710286

Encompass

Catalog No. 71-028-6


Add to cart

Description

Description

The His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). It is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. The His tag specific antibody is used to facilitate detection or coimmunopreciptation of tagged proteins.

710286 detects 6x-His Epitope Tag in Tag samples. 710286 has been successfully used in WB procedures. The immunogen for 710286 is synthetic construct corresponding to amino acids HHHHHH.

Applications:

Western Blotting

Specifications

Specifications

6x-His Epitope Tag
Unconjugated
Purified
Synthetic construct corresponding to amino acids HHHHHH
Protein A
Primary
Tag
21HCLC
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit