Learn More
6x-His Tag Recombinant Polyclonal Antibody (21HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Supplier: Thermo Scientific 710286
Description
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
6x-His Tag Recombinant Polyclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Specifications
6x-His Tag | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Rabbit | |
Protein A | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Immunocytochemistry, Western Blot | |
21HCLC | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
Synthetic construct corresponding to amino acids HHHHHH | |
100 μg | |
Primary | |
Antibody | |
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.