Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

A2ML1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524660

Catalog No. PIPA524660


Add to cart

Description

Description

A2ML1 is a new member of the alpha 2-macroglobulin protease inhibitor family, and shares most the characteristics of this family. It is the first family member detected in the epidermis and may play an important role during desquamation by inhibiting extracellular proteases.
Specifications

Specifications

A2ML1
Polyclonal
PBS with 0.09% sodium azide
A8K2U0, Q3UU35
CPAMD9, C3 and PZP-like, alpha-2-macroglobulin domain containing 9, alpha-2-macroglobulin-like protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
144568, 232400
Flow Cytometry, Western Blot
Unconjugated
A2ML1
Liquid
A2ML1
KLH conjugated synthetic peptide between 345-373 amino acids from the N-terminal region of human A2ML1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit