Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AADAT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523670

Catalog No. PIPA523670

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
Specifications

Specifications

AADAT
Polyclonal
PBS with 0.09% sodium azide
Q8N5Z0
KAT2, KATII, 2-aminoadipate aminotransferase, 2-aminoadipate transaminase, KAT/AadAT, L kynurenine/alpha aminoadipate aminotransferase, alpha-aminoadipate aminotransferase, kynurenine aminotransferase II, kynurenine--oxoglutarate aminotransferase II, kynurenine--oxoglutarate
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51166
Western Blot
Unconjugated
AADAT
Liquid
AADAT
KLH conjugated synthetic peptide between 196-224 amino acids from the central region of human AADAT
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit