Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AA5 Virus VP3 Rabbit anti-Virus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522817

Catalog No. PIPA522817


Add to cart

Description

Description

Adeno-associated virus (AAV) is a small virus which infects humans and some other primate species. AAV is not currently known to cause disease. The virus causes a very mild immune response, lending further support to its apparent lack of pathogenicity.
Specifications

Specifications

AA5 Virus VP3
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
HADV-5; AdV5; AV5; Adeno-associated virus 5; AAV-5 VP3; Adeno-associated 5 Virus VP3; AAV; Lentivirus
Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit