Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ABCC10, Polyclonal, Novus Biologicals
Goat Polyclonal Antibody
Manufacturer: Novus Biologicals NBP120943
Description
ABCC10 Polyclonal antibody specifically detects ABCC10 in Human samples. It is validated for Peptide ELISA.Specifications
ABCC10 | |
0.5 mg/mL | |
Peptide ELISA Detection limit 1:32000 | |
Q5T3U5 | |
ATP-binding cassette sub-family C member 10, ATP-binding cassette, sub-family C (CFTR/MRP), member 10, MRP7SIMRP7EST182763, multidrug resistance-associated protein 7 | |
Goat | |
IgG | |
0.1 mg | |
Store at -20C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Affinity Purified | |
ABCC10 | |
Peptide with sequence C-QLKAENSSQEAQP corresponding to internal region according to NP_258261.2. | |
Immunogen affinity purified | |
RUO | |
Primary | |
89845 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only