Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
MRP2 Mouse anti-Human, Clone: M2III-6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135676
Catalog No. 01-671-082
Description
Description
ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. This protein is expressed in the canalicular (apical) part of the hepatocyte and functions in biliary transport. Substrates include anticancer drugs such as vinblastine; therefore, this protein appears to contribute to drug resistance in mammalian cells. Several different mutations in this gene have been observed in patients with Dubin-Johnson syndrome (DJS), an autosomal recessive disorder characterized by conjugated hyperbilirubinemia.
Specifications
Specifications
MRP2
Monoclonal
Unconjugated
ABCC2
Liquid
ABCC2
Fusion protein of cMOAB/MRP2.
1 mL
4° C
Primary
1244
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
M2III-6
tissue culture supernatant with 0.7% BSA and 0.1% sodium azide
Q92887
MRP2
Mouse
IgG2a
RUO
Antibody
Monoclonal
Human