Learn More
ABCF1 Mouse anti-Human, Clone: 1B4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518335
Catalog No. PIMA518335
Add to cart
Description
Description
Peptide Sequence: GEMRKNHRLK IGFFNQQYAE QLRMEETPTE YLQRGFNLPY QDARKCLGRF GLESHAHTIQ ICKLSGGQKA RVVFAELACR EPDVLILDEP TNNLDIES
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the GCN20 subfamily. Unlike other members of the superfamily, this protein lacks the transmembrane domains which are characteristic of most ABC transporters. This protein may be regulated by tumor necrosis factor-alpha and play a role in enhancement of protein synthesis and the inflammation process.
Specifications
Specifications
ABCF1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8NE71
ABC27, ABC50, ATP-binding cassette 50 (TNF-alpha stimulated), ATP-binding cassette sub-family F member 1, TNF-alpha-stimulated ABC protein, TNFalpha-inducible ATP-binding protein
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23
ELISA, Immunocytochemistry, Immunofluorescence
1B4
Unconjugated
ABCF1
Liquid
ABCF1
ABCF1 (NP_001081, 642 a.a. ∽ 739 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human