Learn More
ABCG1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA513462
Catalog No. PIPA513462
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with bovine, chicken, canine, equine, hamster, mouse, porcine, non-human primate, rat, rabbit and Xenopus, yeast and zebrafish based on sequence homology.
ABCG1 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the White subfamily. It is involved in macrophage cholesterol and phospholipids transport, and may regulate cellular lipid homeostasis in other cell types.
Specifications
Specifications
ABCG1
Polyclonal
Unconjugated
ABCG1
Liquid
ABCG1
KLH conjugated synthetic peptide between 168-195 amino acids from the N-terminal region of human ABCG1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2mg/mL
PBS with 0.09% sodium azide
P45844, Q64343
ATP-binding cassette sub-family G member 1; ATP-binding cassette transporter 8; White protein homolog; WHT1; ABC8; ABCG1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11307, 9619