Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

abhydrolase domain containing 10, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant ABHD10.

Manufacturer:  Abnova Corporation H00055347A01

Catalog No. 89-004-663


Add to cart

Description

Description

Sequence: MAVARLAAVAAWVPCRSWGWAAVPFGPHRGLSVLLARIPQRAPRWLPACRQKTSLSFLNRPDLPNLAYKKLKGKSPGIIFIPGYLSYMNGTKALAIEEFCKSLGHACIRFDYSGVGSSDGNSEESTLGKWRKDVLSIIDDLADGPQILVGSSLGGWLMLHAAIARPEKVVALIGVATAADTLVTKFNQLPVELKKEVEMKGVWSMPSKYSEEGVYNVQYSFIKEAEHHCLLHSPIPVNCPIRLLHGMKDDIVPWHTSMQVADRVLSTDVDVILRKHSDHRMREKADIQLLVYTIDDLIDKLSTIVN
Specifications

Specifications

abhydrolase domain containing 10
Unconjugated
50% glycerol
BC014516
FLJ11342
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
55347
ELISA, Western Blot
Mouse polyclonal antibody raised against a full-length recombinant ABHD10.
ABHD10
Serum
ABHD10
ABHD10 (AAH14516, 1 a.a. ~ 306 a.a) full-length recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit