Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524492

Catalog No. PIPA524492

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

This gene is upstream of, and in a head-to-head orientation with the gene for the mitochondrial ribosomal protein L34. The predicted protein contains alpha/beta hydrolase fold and secretory lipase domains.
Specifications

Specifications

ABHD8
Polyclonal
PBS with 0.09% sodium azide
Q96I13
abhydrolase domain-containing protein 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79575
Western Blot
Unconjugated
ABHD8
Liquid
ABHD8
KLH conjugated synthetic peptide between 410-439 amino acids from the C-terminal region of human ABHD8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit