Learn More
ABHEB Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526705
Catalog No. PIPA526705
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This antibody is predicted to react with bovine based on sequence homology.
ABHEB has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription.
Specifications
Specifications
ABHEB
Polyclonal
Unconjugated
ABHD14B
Liquid
ABHD14B
KLH conjugated synthetic peptide between 150-179 amino acids from the C-terminal region of human ABHEB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q96IU4
CIB, CCG1-interacting factor B, abhydrolase domain-containing protein 14B, cell cycle gene 1-interacting factor B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84836