Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abl2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527926

Catalog No. PIPA527926


Add to cart

Description

Description

PA5-27926 targets ABL2 in WB applications and shows reactivity with Human samples. The PA5-27926 immunogen is recombit fragment corresponding to a region within amino acids 55 and 470 of Human ABL2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. This gene is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
Specifications

Specifications

ABL2
Polyclonal
Unconjugated
ABL2
Liquid
ABL2
Recombit fragment corresponding to a region within amino acids 55 and 470 of Human ABL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.77 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P42684
ABLL, ARG, Abelson tyrosine-protein kinase 2, abelson-related gene protein, tyrosine-protein kinase ARG
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit