Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABLIM1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ABLIM1 protein.

Manufacturer:  Abnova Corporation H00003983B01P

Catalog No. 89-002-563


Add to cart

Description

Description

This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MFTEGEEMYLQGSTVWHPDCKQSTKTEEKLRAKVDNEILDYKDLAAIPKVKAIYDIERPDLITYEPFYTSGYDDKQERQSLGESPRTLSPTPSAEGYQDVRDRMIHRSTSQGSINSPVYSRHSYTPTTSRSPQHFHRPDQGINIYRKPPIYKQHAALAAQSKSSEDIIKFSKFPAAQAPDPSETPKIETDHWPGPPSFAVVGPDMKRRSSGREEDDEELLRRRQLQEEQLMKLNSGLGQLILKEEMEKESRERSSLLASRYDSPINSASHIPSSKTASLPGYGRNGLHRPVSTDFAQYNSYGDVSGGVRDYQTLPDGHMPAMRMDRGVSMPNMLEPKIFPYEMLMVTNRGRNKILREVDRTRLERHLAPEVFREIFGMSIQEFDRLPLWRRNDMKKKAKLF
Specifications

Specifications

ABLIM1
Unconjugated
In 1X PBS, pH 7.4
BC002448.2
ABLIM|DKFZp781D0148|FLJ14564|KIAA0059|LIMAB1|LIMATIN|MGC1224
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human ABLIM1 protein.
ABLIM1
Purified
ABLIM1
ABLIM1 (AAH02448.1, 1 a.a. ~ 401 a.a) full-length human protein.
RUO
Primary
3983
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit