Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAD11 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112907

Catalog No. PA112907


Add to cart

Description

Description

ACAD11 was isolated origainlly from rat liver peroxisomes. 34 known peroxisomal proteins have been identified in addition to several mitochondrial and microsomal proteins. ACAD11 is a bi-functional protein consisting of an aminoglycoside phosphotransferase-domain and an acyl-CoA dehydrogenase domain.
Specifications

Specifications

ACAD11
Polyclonal
Unconjugated
ACAD11
Liquid
ACAD11
Synthetic peptide corresponding to the human ACAD11 protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.08% sodium azide
Q709F0
FLJ12592, MGC150619
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
84129
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit