Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

acyl-Coenzyme A dehydrogenase family, member 8, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant ACAD8.

Manufacturer:  Abnova Corporation H00027034A01

Catalog No. 89-004-594


Add to cart

Description

Description

Isobutyryl-CoA dehydrogenase (EC 1.3.99), encoded by the ACAD8 gene, catalyzes the third step of the degradation of the branched chain amino acid valine (Nguyen et al., 2002 [PubMed 12359132]).[supplied by OMIM

Sequence: GEPLASNQYLQFTLADMATRLVAARLMVRNAAVALQEERKDAVALCSMAKLFATDECFAICNQALQMHGGYGYLKDYAVQQYVRDSRVHQILEGSNEVMRILISRSLLQE
Specifications

Specifications

acyl-Coenzyme A dehydrogenase family, member 8
Unconjugated
50% glycerol
NM_014384
ACAD-8|FLJ22590
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
27034
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant ACAD8.
ACAD8
Serum
ACAD8
ACAD8 (NP_055199, 306 a.a. ~ 415 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit