Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ACAD8, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132224

Catalog No. NBP132224


Add to cart

Description

Description

ACAD8 Polyclonal antibody specifically detects ACAD8 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ACAD8
0.29mg/mL
Western Blot 1:500-1:3000
Q9UKU7
ACAD-8, Activator-recruited cofactor 42 kDa component, Acyl-CoA dehydrogenase family member 8, acyl-CoA dehydrogenase family, member 8, acyl-Coenzyme A dehydrogenase family, member 8, ARC42, EC 1.3.99, EC 1.3.99.-, FLJ22590, IBD, isobutyryl-CoA dehydrogenase, mitochondrial
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Possible cross-reactivity based on sequence identity: Mouse (89%), Pig (91%), Xenopus laevis (82%), Zebrafish (82%), Bovine (88%).
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
ACAD8
Recombinant protein encompassing a sequence within the center region of human ACAD8. The exact sequence is proprietary.
45 kDa
0.1 ml
Core ESC Like Genes, Stem Cell Markers
Primary
27034
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only