Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAD8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526497

Catalog No. PIPA526497

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ACAD8 is a member of the acyl-CoA dehydrogenase family of enzymes that catalyze the dehydrogenation of acyl-CoA derivatives in the metabolism of fatty acids or branch chained amino acids. This protein is a mitochondrial enzyme that functions in catabolism of the branched-chain amino acid valine.
Specifications

Specifications

ACAD8
Polyclonal
PBS with 0.09% sodium azide
Q9UKU7, Q9D7B6
ACAD-8, ARC42, activator-recruited cofactor 42 kDa component, acyl-Coenzyme A dehydrogenase family, member 8, isobutyryl-CoA dehydrogenase, mitochondrial
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27034, 66948
Western Blot
Unconjugated
ACAD8
Liquid
ACAD8
KLH conjugated synthetic peptide between 118-145 amino acids from the central region of human ACAD8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit