Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADM Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527201

Catalog No. PIPA527201


Add to cart

Description

Description

PA5-27201 targets ACADM in IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-27201 immunogen is a recombit fragment corresponding to a region within amino acids 67 and 421 of ACADM. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

ACADM
Polyclonal
Unconjugated
ACADM
Liquid
ACADM
Recombit fragment contain a sequence corresponding to a region within amino acids 67 and 421 of ACADM
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.55 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P11310, P45952, P08503
ACAD1, MCAD, MCADH, acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain, medium-chain specific acyl-CoA dehydrogenase, mitochondrial
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11364, 24158, 34
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit