Learn More
ACAT2 Monoclonal Antibody (4A5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA518338
Description
Peptide Sequence: MNAGSDPVVI VSAARTIIGS FNGALAAVPV QDLGSTVIKE VLKRATVAPE DVSEVIFGHV LAAGCGQNPV RQASVGAGIP YSVPAWSCQM ICGSGLKAVC LAVQSIGIGD SSIVVAGGME NMSKAPHLAY LRTGVKIGEM PLTDSILCDG LTDAFHNCHM GITAENVAKK WQVSREDQDK VAVLSQNRTE NAQKAGHFDK EIVPVLVSTR RGLIEVKTDE FPRHGSNIEA MSKLKPYFLT DGTGTVTPAN ASGINDGAAA VVLMKKSEAD KRGLTPLARI VSWSQVGVEP SIMGIGPIPA IKQAVTKAGW SLEDVDIFEI NEAFAAVSAA IVKELGLNPE KVNIEGGAIA LGHPLGASGC RILVTLLHTL ERMGRSRGVA ALCIGGGMGI AMCVQRE
ACAT2 Monoclonal antibody specifically detects ACAT2 in Human samples. It is validated for ELISA, Western Blot.Specifications
ACAT2 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9BWD1 | |
Acat2 | |
ACAT2 (AAH00408, 1 a.a. ∽ 397 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4A5 | |
Unconjugated | |
Acat2 | |
Ab2-076; Acat2; Acat3; acetoacetyl Coenzyme A thiolase; acetyl-CoA acetyltransferase 2; acetyl-CoA acetyltransferase, cytosolic; Acetyl-CoA transferase-like protein; acetyl-Coenzyme A acetyltransferase 2; acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase); acetyl-Coenzyme A acetyltransferase 3; ACTL; AE1; AW742799; BND3; CD233; cytosolic acetoacetyl-CoA thiolase; DI; EMPB3; EPB3; FR; RTA1A; SW; t-complex protein 1, related sequence 1; Tcp1-rs1; Tcp-1x; WD; WD1; WR | |
Mouse | |
Affinity chromatography | |
RUO | |
39 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.